Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-5-methyl-2-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A101329-1G A101329-5G A101329-100MG A101329-250MG A101329-1G
103795-910EA 77 USD InStock
103795-910 103821-596 76780-488 76780-490 77282-700
4-Methoxy-5-methyl-2-nitroaniline 98%
4-Methoxy-5-methyl-2-nitroaniline
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₃
MW: 182.18 g/mol
Boiling Pt: 355.1±37.0 °C (760 mm Hg)
Melting Pt: 117.5…118.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00031126
CAS Number: 55730-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty