Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Anisohydrazide 98%

Supplier: AMBEED, INC
Warning

A149932-5G A149932-100G A149932-500G A149932-25G
103808-732EA 445 USD InStock
103808-732 103842-228 77190-764 77190-766 77317-748
p-Anisohydrazide 98%
p-Anisohydrazide
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₂
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00017073
CAS Number: 3290-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty