Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-1H-indole-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A189723-1G A189723-5G A189723-100MG A189723-250MG A189723-25G A189723-1G A189723-5G
103816-218EA 41 USD InStock
103816-218 103846-812 77274-396 77274-398 77274-400 77302-184 77302-186
4-Methyl-1H-indole-2-carboxylic acid 97%
4-Methyl-1H-indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MW: 175.1800 g/mol
Boiling Pt: 419.9 °C (760 mm Hg)
Melting Pt: 220…221 °C
Flash Pt: 207.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02664476
CAS Number: 18474-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty