Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A500108-1G A500108-5G A500108-250MG
103828-288EA 51 USD InStock
103828-288 103839-376 76843-336 76843-338 77034-496
4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid 95%
4-Methyl-3-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)benzoic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₉BO₄
Melting Pt: 237…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09261433
CAS Number: 515131-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty