Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylbenzamidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A161764-1G A161764-5G A161764-10G A161764-250MG A161764-1G A161764-25G A161764-5G
103805-532EA 19 USD InStock
103805-532 103843-774 77191-100 77191-102 77319-780 77319-782 77319-784
4-Methylbenzamidine hydrochloride 97%
4-Methylbenzamidine hydrochloride
Precaución: Inert atmosphere, room temperature


Formula: C₈H₁₁ClN₂
MW: 170.64
Storage Temperature: Ambient
MDL Number: MFCD00040183
CAS Number: 6326-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty