Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phenyl-p-toluidine 97%

Supplier: AMBEED, INC
Warning

A140855-100G A140855-10G A140855-500G A140855-25G A140855-5G
76719-350EA 37 USD InStock
76719-350 76719-352 76719-354 77316-026 77316-028
N-Phenyl-p-toluidine 97%
N-Phenyl-p-toluidine
Precaución: for Research Use Only


Formula: C₁₃H₁₃N
Boiling Pt: 334 °C (760 mm Hg)
Melting Pt: 90 °C
Flash Pt: 154.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00092921
CAS Number: 620-84-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty