Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylpiperazin-2-one 97%

Supplier: AMBEED, INC
Warning

A117289-1G A117289-25G A117289-1G A117289-10G A117289-250MG
103798-862EA 21 USD InStock
103798-862 103824-736 77026-188 77297-530 77880-945 77880-946
4-Methylpiperazin-2-one 97%
4-Methylpiperazin-2-one
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O
Boiling Pt: 261.3 °C (760 mm Hg)
Melting Pt: 85…89 °C
Flash Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11870124
CAS Number: 34770-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty