Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylthiazole-5-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A380806-1G A380806-5G A380806-10G A380806-1G A380806-100G
103826-680EA 5 USD InStock
103826-680 103837-714 76841-034 77290-978 77306-918 77748-767
4-Methylthiazole-5-carboxylic acid 98%
4-Methylthiazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₅H₅NO₂S
Boiling Pt: 303.6 °C (760 mm Hg)
Melting Pt: 287 °C
Flash Pt: 137.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00626872
CAS Number: 20485-41-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty