Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Piperidin-4-ylmorpholine 97%

Supplier: AMBEED, INC
Warning

A110031-1G A110031-5G A110031-100G A110031-1G A110031-5G A110031-10G
103797-608EA 5 USD InStock
103797-608 103823-416 77093-956 77310-222 77310-224 77565-940
4-Piperidin-4-ylmorpholine 97%
4-Piperidin-4-ylmorpholine
Precaución: For research use only.


Formula: C₉H₁₈N₂O
Boiling Pt: 256.3 °C (760 mm Hg)
Melting Pt: 40…43 °C
Flash Pt: 108.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03274733
CAS Number: 53617-35-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty