Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophthalic acid 98%

Supplier: AMBEED, INC
Warning

A175525-5G A175525-1000G A175525-100G A175525-500G A175525-5G
103845-362EA 5 USD InStock
103845-362 77236-398 77236-400 77236-402 77301-378
4-Nitrophthalic acid 98%
4-Nitrophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅NO₆
Boiling Pt: 459.5 °C (760 mm Hg)
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00007252
CAS Number: 610-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty