Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrosalicylic acid 97%

Supplier: AMBEED, INC
Warning

A242989-1G A242989-5G A242989-1G A242989-5G A242989-10G
103811-496EA 7 USD InStock
103811-496 103851-320 77090-512 77090-514 77140-326
4-Nitrosalicylic acid 97%
4-Nitrosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Boiling Pt: 389 °C (760 mm Hg)
Melting Pt: 235…239 °C
Flash Pt: 179.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00071505
CAS Number: 619-19-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty