Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Oxazole-4-carbaldehyde 96%

Supplier: AMBEED, INC
Warning

A252394-1G A252394-5G A252394-100MG A252394-250MG A252394-5G
103812-040EA 39 USD InStock
103812-040 103851-884 77014-020 77014-022 77304-582
1,3-Oxazole-4-carbaldehyde 96%
1,3-Oxazole-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₄H₃NO₂
MW: 97.07 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179512
CAS Number: 118994-84-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty