Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Oxocyclohexanecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A158530-1G A158530-5G A158530-100G A158530-10G A158530-250MG A158530-500G A158530-1G A158530-25G A158530-5G
103805-136EA 9 USD InStock
103805-136 103843-348 77286-348 77286-350 77286-352 77286-354 77300-334 77319-210 77319-212
4-Oxocyclohexanecarboxylic acid 97%
4-Oxocyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MW: 142.16 g/mol
Boiling Pt: 309.7 °C (760 mm Hg)
Melting Pt: 67…71 °C
Flash Pt: 155.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00102035
CAS Number: 874-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty