Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentynoic acid 98%

Supplier: AMBEED, INC
Danger

A391230-5G A391230-1G A391230-5G A391230-10G
103826-878EA 10 USD InStock
103826-878 103837-918 76841-308 76841-310 77016-182
4-Pentynoic acid 98%
4-Pentynoic acid


Formula: C₅H₆O₂
Boiling Pt: 199.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00004407
CAS Number: 6089-09-4
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty