Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Piperidinyl)aniline 97%

Supplier: AMBEED, INC
Warning

A426308-1G A426308-5G
103827-470EA 13 USD InStock
103827-470 103838-520 76842-174 76842-176 77102-194
4-(1-Piperidinyl)aniline 97%
4-(1-Piperidinyl)aniline
Precaución: for Research Use Only


Formula: C₁₁H₁₆N₂
Boiling Pt: 345.1 °C (760 mm Hg)
Melting Pt: 26…29 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051688
CAS Number: 2359-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty