Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4'-Bipiperidine 98%

Supplier: AMBEED, INC
Warning

A197582-1G A197582-5G A197582-10G A197582-25G A197582-5G A197582-1G
103816-944EA 7 USD InStock
103816-944 103847-564 76691-092 76691-094 77085-578 77302-556
1,4'-Bipiperidine 98%
1,4'-Bipiperidine
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Boiling Pt: 251.4 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 99.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00006475
CAS Number: 4897-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty