Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Propoxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A173508-1G A173508-5G A173508-5G A173508-10G A173508-1G A173508-25G
103806-822EA 9 USD InStock
103806-822 103845-122 77082-364 77096-556 77301-276 77301-278
4-Propoxyphenylboronic acid 97%
4-Propoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427189
CAS Number: 186497-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty