Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isonicotinaldehyde 98%

Supplier: AMBEED, INC
Warning

A105755-5G A105755-100G A105755-10G A105755-25G A105755-500G A105755-5G
103822-672EA 5 USD InStock
103822-672 77107-570 77107-572 77107-574 77107-576 77309-538
Isonicotinaldehyde 98%
Isonicotinaldehyde
Precaución: for Research Use Only


Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 71…73 °C (10 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00006425
CAS Number: 872-85-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty