Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Vinylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A125275-1G A125275-100G A125275-10G A125275-25G A125275-1G A125275-5G
103800-660EA 9 USD InStock
103800-660 77258-236 77258-238 77258-240 77298-040 77312-812
4-Vinylbenzoic acid 98%
4-Vinylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 282
Melting Pt: 115
Flash Pt: 132.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty