Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Vinylphenol 10% (w/w) in propylene glycol 10 wt.%

Supplier: AMBEED, INC
Warning

A302673-1G A302673-100G A302673-10G A302673-500G A302673-25G A302673-5G A302673-1G
103814-346EA 7 USD InStock
103814-346 103835-504 76772-670 76772-672 76772-674 76838-186 76838-188 77305-854
4-Vinylphenol 10% (w/w) in propylene glycol 10 wt.%
4-Vinylphenol
Precaución: for Research Use Only


Formula: C₈H₈O
Boiling Pt: 206.2 °C (760 mm Hg)
Flash Pt: 92.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017593
CAS Number: 2628-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty