Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(1-Hydroxy-2-(2,4,5-trifluorophenyl)ethylidene)-2,2-dimethyl-1,3-dioxane-4,6-dione 97%

Supplier: AMBEED, INC
Warning

A408083-1G A408083-5G A408083-1G A408083-5G A408083-100G A408083-10G
103827-176EA 18 USD InStock
103827-176 103838-220 77307-122 77307-124 77878-156 77878-157
5-(1-Hydroxy-2-(2,4,5-trifluorophenyl)ethylidene)-2,2-dimethyl-1,3-dioxane-4,6-dione 97%
5-(1-Hydroxy-2-(2,4,5-trifluorophenyl)ethylidene)-2,2-dimethyl-1,3-dioxane-4,6-dione
Precaución: for Research Use Only


Formula: C₁₄H₁₁F₃O₅
Boiling Pt: 489.0±45.0 °C (760 mm Hg)
Melting Pt: 117 °C
Flash Pt: 249.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12407134
CAS Number: 764667-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty