Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminopyridine-3-boronic acid, pinacol ester 95%

Supplier: AMBEED, INC
Warning

A218050-1G A218050-5G A218050-10G A218050-250MG A218050-5G
103818-684EA 26 USD InStock
103818-684 103849-416 76785-884 76785-886 77088-072
5-Aminopyridine-3-boronic acid, pinacol ester 95%
5-Aminopyridine-3-boronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₁₇BN₂O₂
MW: 220.076 g/mol
Melting Pt: 194…195 °C
Storage Temperature: Freezer
MDL Number: MFCD09260489
CAS Number: 1073354-99-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty