Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Bromophenyl)-1H-tetrazole 98%

Supplier: AMBEED, INC
Danger

A165748-1G A165748-5G A165748-10G A165748-5G
103805-990EA 45 USD InStock
103805-990 103844-246 77138-306 77320-468
5-(4-Bromophenyl)-1H-tetrazole 98%
5-(4-Bromophenyl)-1H-tetrazole
Precaución: for Research Use Only


Formula: C₇H₅BrN₄
Boiling Pt: 374.6 °C (760 mm Hg)
Melting Pt: 260 °C
Flash Pt: 180.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00991874
CAS Number: 50907-23-8
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty