Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-2,4-dichloropyrimidine 98%

Supplier: AMBEED, INC
Warning

A140504-1G A140504-100MG A140504-250MG A140504-25G A140504-1G A140504-5G
103802-880EA 20 USD InStock
103802-880 76811-744 76811-746 76811-748 77299-150 77299-152
5-(Benzyloxy)-2,4-dichloropyrimidine 98%
5-(Benzyloxy)-2,4-dichloropyrimidine
Precaución: for Research Use Only


Formula: C₁₁H₈Cl₂N₂O
MW: 255.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658924
CAS Number: 91183-17-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty