Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid 96%

Supplier: AMBEED, INC
Warning

A252503-1G A252503-5G A252503-250MG A252503-1G
103812-046EA 123 USD InStock
103812-046 103851-890 76826-996 77304-584
5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid 96%
5-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₅
MW: 246.216 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD08531038
CAS Number: 1219-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty