Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)pentan-1-ol 95%

Supplier: AMBEED, INC
Warning

A101007-1G A101007-5G A101007-10G A101007-25G A101007-5G A101007-1G
103795-750EA 10 USD InStock
103795-750 103821-442 76765-154 76765-156 77282-638 77296-948
5-(Benzyloxy)pentan-1-ol 95%
5-(Benzyloxy)pentan-1-ol
Precaución: for Research Use Only


Formula: C₁₂H₁₈O₂
Boiling Pt: 296.8 °C (760 mm Hg)
Flash Pt: 119.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00216583
CAS Number: 4541-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty