Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)pyridin-2-amine 97%

Supplier: AMBEED, INC
Warning

A132697-1G A132697-250MG A132697-5G
103801-758EA 16 USD InStock
103801-758 76972-156 77314-346
5-(Benzyloxy)pyridin-2-amine 97%
5-(Benzyloxy)pyridin-2-amine
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂O
Flash Pt: 180.289 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11135746
CAS Number: 96166-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty