Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chloromethyl)-2-methoxypyridine 95%

Supplier: AMBEED, INC
Danger

A296992-1G A296992-5G A296992-100MG A296992-250MG A296992-1G
103814-118EA 186 USD InStock
103814-118 103835-260 76827-778 76827-780 76837-944
5-(Chloromethyl)-2-methoxypyridine 95%
5-(Chloromethyl)-2-methoxypyridine
Precaución: for Research Use Only


Formula: C₇H₈ClNO
Flash Pt: 96.6 °C
Storage Temperature: Freezer
MDL Number: MFCD10697570
CAS Number: 101990-70-9
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty