Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Hydroxymethyl)dihydrofuran-2(3H)-one ≥95%

Supplier: AMBEED, INC
Warning

A153062-1G A153062-5G A153062-250MG
103804-462EA 22 USD InStock
103804-462 77871-754 77871-753
5-(Hydroxymethyl)dihydrofuran-2(3H)-one ≥95%
5-(Hydroxymethyl)dihydrofuran-2(3H)-one
Precaución: Sealed in dry, store in freezer, under –20 °C.


Formula: C₅H₈O₃
MW: 116.12 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00506240
CAS Number: 10374-51-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty