Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Hydroxymethyl)nicotinonitrile 97%

Supplier: AMBEED, INC
Danger

A116541-1G A116541-100MG A116541-10G A116541-250MG A116541-5G A116541-1G
103798-752EA 79 USD InStock
103798-752 77136-292 77136-294 77136-296 77136-298 77311-424
5-(Hydroxymethyl)nicotinonitrile 97%
5-(Hydroxymethyl)nicotinonitrile
Precaución: for Research Use Only


Formula: C₇H₆N₂O
Boiling Pt: 315 °C (760 mm Hg)
Flash Pt: 144.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367899
CAS Number: 135124-71-9
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty