Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethoxy)-1H-indole 97%

Supplier: AMBEED, INC
Warning

A225697-1G A225697-5G A225697-100MG A225697-250MG A225697-1G A225697-5G
103819-188EA 98 USD InStock
103819-188 103850-088 76874-100 76874-102 77303-746 77303-748
5-(Trifluoromethoxy)-1H-indole 97%
5-(Trifluoromethoxy)-1H-indole
Precaución: for Research Use Only


Formula: C₉H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09954768
CAS Number: 262593-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty