Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Trifluoromethyl-1H-indole-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A249324-1G A249324-5G A249324-1G A249324-5G A249324-100MG A249324-250MG
103811-866EA 144 USD InStock
103811-866 103851-702 77091-048 77091-050 77113-414 77113-416
5-Trifluoromethyl-1H-indole-2-carboxylic acid 95%
5-Trifluoromethyl-1H-indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
Boiling Pt: 402.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03095229
CAS Number: 496946-78-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty