Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: AMBEED, INC
Danger

A252522-1G A252522-5G A252522-10G A252522-250MG A252522-1G A252522-5G
103812-048EA 20 USD InStock
103812-048 103851-892 76786-832 76786-834 77091-306 77091-308
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 98%
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
Boiling Pt: 270.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06659680
CAS Number: 1036027-54-9
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty