Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 98%

Supplier: AMBEED, INC
Warning

A130488-1G A130488-100MG A130488-10G A130488-250MG A130488-1G A130488-5G
103801-432EA 91 USD InStock
103801-432 77094-942 77094-944 77094-946 77313-870 77313-872
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one 98%
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only


Formula: C₁₀H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD07772122
CAS Number: 150969-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty