Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-(trifluoromethyl)pyrazine 97%

Supplier: AMBEED, INC
Warning

A224749-1G A224749-5G A224749-250MG A224749-1G A224749-5G A224749-100MG
103819-130EA 38 USD InStock
103819-130 103850-022 76786-106 77088-686 77088-688 77667-870
2-Amino-5-(trifluoromethyl)pyrazine 97%
2-Amino-5-(trifluoromethyl)yrazine
Precaución: for Research Use Only


Formula: C₅H₄F₃N₃
Boiling Pt: 226.204 °C (760 mm Hg)
Melting Pt: 131…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697797
CAS Number: 69816-38-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty