Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Trifluoromethyl)uracil 97%

Supplier: AMBEED, INC
Warning

A159127-1G A159127-5G A159127-100G A159127-10G A159127-1G A159127-25G A159127-5G
103805-226EA 6 USD InStock
103805-226 103843-442 76973-458 76973-460 76973-462 77300-388 77319-316 77319-318
5-(Trifluoromethyl)uracil 97%
5-(Trifluoromethyl)uracil
Precaución: for Research Use Only


Formula: C₅H₃F₃N₂O₂
Melting Pt: 245…246 °C
Storage Temperature: Freezer
MDL Number: MFCD00006024
CAS Number: 54-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty