Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine 95%

Supplier: AMBEED, INC
Warning

A143453-1G A143453-10G A143453-250MG A143453-1G A143453-5G
103803-286EA 23 USD InStock
103803-286 76767-910 76767-912 77299-342 77316-556
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine 95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine
Precaución: for Research Use Only


Formula: C₆H₉N₃
Boiling Pt: 322.8 °C (760 mm Hg)
Flash Pt: 149.046 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09701264
CAS Number: 91476-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty