Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Difluoroindole 98%

Supplier: AMBEED, INC
Warning

A100948-1G A100948-5G A100948-100MG A100948-10G A100948-250MG A100948-25G A100948-1G A100948-5G
103795-716EA 31 USD InStock
103795-716 103821-406 77270-176 77270-178 77270-180 77270-182 77296-924 77296-926
5,6-Difluoroindole 98%
5,6-Difluoroindole
Precaución: for Research Use Only


Formula: C₈H₅F₂N
Boiling Pt: 264.5 °C (760 mm Hg)
Flash Pt: 113.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01075212
CAS Number: 169674-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty