Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5,6-dimethylbenzothiazole 97%

Supplier: AMBEED, INC
Warning

A182300-1G A182300-5G A182300-1G A182300-100G A182300-10G A182300-250MG
103807-694EA 15 USD InStock
103807-694 103846-064 77083-604 77248-308 77248-310 77248-312
2-Amino-5,6-dimethylbenzothiazole 97%
2-Amino-5,6-dimethylbenzothiazole
Precaución: for Research Use Only


Formula: C₉H₁₀N₂S
Boiling Pt: 342.1 °C (760 mm Hg)
Melting Pt: 185…189 °C
Flash Pt: 160.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005790
CAS Number: 29927-08-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty