Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Dimethylpyrazin-2-amine 97%

Supplier: AMBEED, INC
Warning

A218090-1G A218090-10G A218090-250MG A218090-1G A218090-100MG
103818-690EA 158 USD InStock
103818-690 76873-856 76873-858 77088-074 77671-946
5,6-Dimethylpyrazin-2-amine 97%
5,6-Dimethylpyrazin-2-amine
Precaución: for Research Use Only


Formula: C₆H₉N₃
Boiling Pt: 253.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09835026
CAS Number: 6294-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty