Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,7-Dichloroimidazo[1,2-a]pyrimidine 97%

Supplier: AMBEED, INC
Warning

A167268-250MG A167268-1G
103806-154EA 226 USD InStock
103806-154 76768-920 77081-420
5,7-Dichloroimidazo[1,2-a]pyrimidine 97%
5,7-Dichloroimidazo[1,2-a]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₃Cl₂N₃
MW: 188.01 g/mol
Melting Pt: 257…259 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06858696
CAS Number: 57473-32-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty