Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,8-Dibromoquinoxaline 98%

Supplier: AMBEED, INC
Danger

A516533-100MG A516533-250MG A516533-1G A516533-5G
103828-454EA 32 USD InStock
103828-454 103839-560 76830-338 76830-340 76843-548 76843-550
5,8-Dibromoquinoxaline 98%
5,8-Dibromoquinoxaline
Precaución: for Research Use Only


Formula: C₈H₄Br₂N₂
MW: 287.93 g/mol
Boiling Pt: 352.5±37.0 °C (760 mm Hg)
Melting Pt: 229 °C
Flash Pt: 167 °C
Storage Temperature: Ambient
MDL Number: MFCD18448005
CAS Number: 148231-12-3
UN: 2923
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty