Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,8-Dimethoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-amine 98%

Supplier: AMBEED, INC
Warning

A124871-1G A124871-100G A124871-10G A124871-250MG A124871-25G A124871-500G A124871-1G A124871-5G
103800-586EA 6 USD InStock
103800-586 76870-438 76870-440 76870-442 76870-444 76870-446 77312-728 77312-730
5,8-Dimethoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-amine 98%
5,8-Dimethoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₇H₉N₅O₂
Melting Pt: 201…203 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17168561
CAS Number: 219715-62-5
Order Now
Specification Test Results
Loss on drying ≤ 10%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty