Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-1-naphthoic acid 98%

Supplier: AMBEED, INC
Warning

A117650-1G A117650-100MG A117650-250MG A117650-5G
103798-918EA 195 USD InStock
103798-918 76766-278 76766-280 77876-862
5-Amino-1-naphthoic acid 98%
5-Amino-1-naphthoic acid
Precaución: for Research Use Only


Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Boiling Pt: 447.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00578901
CAS Number: 32018-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty