Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-6-bromo-2,3-dihydro-1H-inden-1-one 96%

Supplier: AMBEED, INC
Warning

A109270-1G A109270-100MG A109270-250MG A109270-5G
103797-480EA 85 USD InStock
103797-480 103823-274 77245-600 77245-602 77283-848
5-Amino-6-bromo-2,3-dihydro-1H-inden-1-one 96%
5-Amino-6-bromo-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only


Formula: C₉H₈BrNO
Boiling Pt: 376.725 °C (760 mm Hg)
Flash Pt: 181.637 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16877204
CAS Number: 158205-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty