Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminoindan-1-one 98%

Supplier: AMBEED, INC
Warning

A174249-1G A174249-5G A174249-1G A174249-25G A174249-5G A174249-10G A174249-250MG
103806-936EA 28 USD InStock
103806-936 103845-234 77082-530 77082-532 77082-534 77273-816 77273-818
5-Aminoindan-1-one 98%
5-Aminoindan-1-one
Precaución: for Research Use Only


Formula: C₉H₉NO
MW: 147.177 g/mol
Boiling Pt: 344.5 °C (760 mm Hg)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00099466
CAS Number: 3470-54-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty