Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminooxindole 95%

Supplier: AMBEED, INC
Warning

A236188-5G A236188-1G A236188-5G A236188-10G A236188-250MG
103811-078EA 23 USD InStock
103811-078 103850-846 77089-892 77089-894 77249-654 77249-656
5-Aminooxindole 95%
5-Aminooxindole
Precaución: for Research Use Only


Formula: C₈H₈N₂O
Boiling Pt: 416.8 °C (760 mm Hg)
Flash Pt: 205.8 °C
Storage Temperature: Freezer
MDL Number: MFCD02179603
CAS Number: 20876-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty