Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1,3-dihydrobenzoimidazol-2-one 97%

Supplier: AMBEED, INC
Warning

A120349-1G A120349-100G A120349-10G A120349-250MG A120349-25G A120349-1G A120349-5G
103799-920EA 19 USD InStock
103799-920 77161-266 77161-268 77161-270 77161-272 77312-030 77312-032
5-Bromo-1,3-dihydrobenzoimidazol-2-one 97%
5-Bromo-1,3-dihydrobenzoimidazol-2-one
Precaución: for Research Use Only


Formula: C₇H₅BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD01893032
CAS Number: 39513-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty