Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromobenzimidazole 98%

Supplier: AMBEED, INC
Warning

A156257-1G A156257-5G A156257-100G A156257-100MG A156257-10G A156257-250MG A156257-1G A156257-25G A156257-5G
103804-852EA 8 USD InStock
103804-852 103843-054 76768-350 76768-352 76768-354 76768-356 77318-806 77318-808 77318-810
5-Bromobenzimidazole 98%
5-Bromobenzimidazole
Precaución: for Research Use Only


Formula: C₇H₅BrN₂
Melting Pt: 130 °C
Flash Pt: 206.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00160001
CAS Number: 4887-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty