Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1H-indole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A326792-1G A326792-25G A326792-5G A326792-100G A326792-10G A326792-1G
103825-448EA 6 USD InStock
103825-448 103836-400 76839-368 76839-370 76875-952 76875-954 77306-354
5-Bromo-1H-indole-3-carbaldehyde 98%
5-Bromo-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 204…207 °C
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00152016
CAS Number: 877-03-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty